Supelco® PESTANAL® Organic Reference Standard, Pyrethrum Extract
Catalog # 33739-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:8003-34-7
- Description:Pyrethrum extract
- Storage conditions:2...30 °C
- Hazard class description:H301 + H331 - H312 - H410
- Shipping conditions:STD
- MDL No.:MFCD00078611
- M-Clarity quality level:MQ100
- Formula:C₄₃H₅₆O₈
- Flash point:75 °C - closed cup
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P261 - P273 - P280 - P301 + P310 - P302 + P352 + P312 - P304 + P340 + P311