Supelco® PESTANAL® Organic Reference Standard, Pyrazosulfuron-ethyl
Catalog # 46323-100MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:93697-74-6
- Description:Pyrazosulfuron-ethyl
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00274605
- M-Clarity quality level:MQ100
- Synonym(s):Ethyl 5-[(4,6-dimethoxy-2-pyrimidinyl)aminocarbonylaminosulfonyl]-1-methyl-1H-pyrazole-4-carboxylate
- Molecular weight:414,39