Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Prallethrin
  32917-100MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Prallethrin
  32917-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    23031-36-9
  • Description:
    Prallethrin
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H302 - H331 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD03792792
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (RS)-2-Methyl-4-oxo-3-prop-2-ynylcyclopent-2-enyl (1RS,3RS;1RS,3SR)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropanecarboxylate
  • Formula:
    C₁₉H₂₄O₃
  • Molecular weight:
    300,39
  • Storage temperature:
    2...8 °C
  • Flash point:
    139 °C - closed cup

 

 

  • GHS signal: GHS06, GHS09
 : P261 - P264 - P270 - P273 - P301 + P312 - P304 + P340 + P311