Order Entry
Slovakia
ContactUsLinkComponent
Pesticide reference standard, Potassium clavulanate, VETRANAL(TM), analytical standard, Supelco®
Pesticide reference standard, Potassium clavulanate, VETRANAL(TM), analytical standard, Supelco®
Catalog # 33454-100MG
Supplier:  Merck
CAS Number:  
undefined
Pesticide reference standard, Potassium clavulanate, VETRANAL(TM), analytical standard, Supelco®
Catalog # 33454-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    61177-45-5
  • Description:
    Potassium clavulanate, VETRANAL(TM)
  • M-Clarity quality level:
    MQ200
  • Formula:
    C₈H₈NO₅K
  • Storage temperature:
    2...8 °C
  • Description:
    Potassium clavulanate, VETRANAL(TM)

Specifications

About this item

β-lactamase inhibitor is added to amoxicillin to increase its effectiveness.