Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Pirimiphos-Ethyl
  45628-250MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Pirimiphos-Ethyl
  45628-250MG
 :  Merck
 :  

 

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    23505-41-1
  • Description:
    Pirimiphos-ethyl
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H301 - H312 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055295
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₃H₂₄N₃O₃PS
  • Molecular weight:
    333,39
  • Storage temperature:
    2...8 °C
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS06, GHS09
 : P264 - P270 - P273 - P280 - P301 + P310 - P302 + P352 + P312