Supelco® PESTANAL® Organic Reference Standard, Piperophos
46011-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:24151-93-7
- Description:Piperophos
- Storage conditions:2…8 °C
- Hazard class description:H302 - H330 - H410
- Shipping conditions:STD
- MDL No.:MFCD00210346
- M-Clarity quality level:MQ100
- Molecular weight:353,48
- Flash point:>100 °C
- GHS signal: GHS06, GHS09
: P273 - P301 + P312 + P330 - P304 + P340 + P310