Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Penthiopyrad
  32064-25MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Penthiopyrad
  32064-25MG
 :  Merck
 :  

 

  • Pk:
    25 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    183675-82-3
  • Description:
    Penthiopyrad
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD11973641
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    (RS)-N-[2-(1,3-Dimethylbutyl)-3-thienyl]-1-methyl-3-(trifluoromethyl)pyrazole-4-carboxamide
  • Formula:
    C₁₆H₂₀F₃N₃OS
  • Molecular weight:
    359,41
  • Flash point:
    Not applicable

 

 

  • GHS signal: GHS09
 : P273 - P391 - P501