Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Paraquat Dichloride Hydrate
Supelco® PESTANAL® Organic Reference Standard, Paraquat Dichloride Hydrate
Catalog # 36541-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Paraquat Dichloride Hydrate
Catalog # 36541-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    75365-73-0
  • Description:
    Paraquat dichloride hydrate
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H301 - H310 + H330 - H315 - H319 - H335 - H372 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00150001
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1,1′-Dimethyl-4,4′-bipyridinium dichloride hydrate; Gramoxone; Methyl viologen dichloride; Paraquat dichloride
  • Formula:
    C₁₂H₁₄Cl₂N₂ ·xH₂O
  • Molecular weight:
    257,16 (anhydrous basis)
  • Storage temperature:
    2...8 °C

Specifications

About this item

  • GHS signal: GHS06, GHS08, GHS09
Caution: P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310 - P314