Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Naptalam
Supelco® PESTANAL® Organic Reference Standard, Naptalam
Catalog # 33371-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Naptalam
Catalog # 33371-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    132-66-1
  • Description:
    Naptalam
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H412
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00037725
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    N-(1-Naphthyl)phthalamidic acid
  • Melting point:
    185...190 °C
  • Formula:
    C₁₈H₁₃NO₃
  • Molecular weight:
    291,30
  • Flash point:
    Not applicable

Specifications

About this item

Caution: P273 - P501