Supelco® PESTANAL® Organic Reference Standard, Methiocarb Sulfoxide
Catalog # 34177-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:2635-10-1
- Description:Methiocarb sulfoxide
- Storage conditions:2...8 °C
- Hazard class description:H300 - H400
- Shipping conditions:STD
- MDL No.:MFCD00155393
- M-Clarity quality level:MQ100
- Synonym(s):4-Methylsulfinyl-3,5-xylyl-N-methylcarbamate; Mercaptodimethur sulfoxide; Mesurol sulfoxide; Methiocarb breakdown product
- Formula:C₁₁H₁₅NO₃S
- Molecular weight:241,31
- Storage temperature:2…8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P273 - P301 + P310 + P330