Pesticide reference standard, Methiocarb sulfone, analytical standard, Supelco®
Catalog # MET543A
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Analytical standard
- CAS No.:2179-25-1
- Description:Methiocarb sulfone
- Formula:C₁₁H₁₅NO₄S
- Description:Methiocarb sulfone
- Pk Info:50 mg/ampule