Supelco® PESTANAL® Organic Reference Standard, Metalaxyl
32012-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:57837-19-1
- Description:Metalaxyl
- Storage conditions:2...30 °C
- Hazard class description:H302 - H317 - H412
- Shipping conditions:STD
- MDL No.:MFCD00055447
- M-Clarity quality level:MQ100
- Synonym(s):N-(2,6-Dimethylphenyl)-N-(methoxyacetyl)-ᴅʟ-alanine methyl ester
- Melting point:69...73 °C
- Formula:C₁₅H₂₁NO₄
- Molecular weight:279,33
- Flash point:100 °C - closed cup
- GHS signal: GHS07
: P261 - P264 - P273 - P280 - P301 + P312 - P302 + P352