Supelco® PESTANAL® Organic Reference Standard, Mefluidide
Catalog # 54746-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:53780-34-0
- Description:Mefluidide
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD01632752
- M-Clarity quality level:MQ100
- Synonym(s):5′-(1,1,1-Trifluoromethanesulfonamido)acet-2′,4′-xylidide; 5-Acetamido-2,4-dimethyltrifluoromethanesulfonanilide; N-[2,4-Dimethyl-5-[[(trifluoromethyl)sulfonyl]amino]phenyl]acetamide
- Molecular weight:310,29