Supelco® PESTANAL® Organic Reference Standard, Mecoprop-P
Catalog # 36773-250MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:16484-77-8
- Description:Mecoprop-P
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00144037
- M-Clarity quality level:MQ100
- Synonym(s):(R)-2-(4-Chloro-2-methylphenoxy)propionic acid
- Formula:C₁₀H₁₁ClO₃
- Molecular weight:214,65
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS05, GHS07, GHS09