Supelco® PESTANAL® Organic Reference Standard, Mecoprop
Catalog # 36147-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:93-65-2
- Description:Mecoprop
- Storage conditions:2...30 °C
- Hazard class description:H302 - H311 - H315 - H318 - H410
- Shipping conditions:STD
- MDL No.:MFCD00002648
- M-Clarity quality level:MQ100
- Synonym(s):2-(4-Chloro-2-methylphenoxy)propionic acid
- Formula:C₁₀H₁₁ClO₃
- Molecular weight:214,65
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS05, GHS09
Caution: P273 - P280 - P301 + P312 + P330 - P302 + P352 + P312 - P305 + P351 + P338 + P310