Supelco® PESTANAL® Organic Reference Standard, Isocarbophos
37901-100MG-R
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:24353-61-5
- Description:Isocarbophos
- Storage conditions:2...8 °C
- Hazard class description:H300 - H311
- Shipping conditions:STD
- MDL No.:MFCD01684421
- M-Clarity quality level:MQ100
- Synonym(s):O-Methyl O-[2-(isopropoxycarbonyl)phenyl] phosphoramidothioate
- Formula:C₁₁H₁₆NO₄PS
- Molecular weight:289,29
- Storage temperature:2...8 °C
- Flash point:Not applicable
- GHS signal: GHS06
: P264 - P270 - P280 - P301 + P310 - P302 + P352 + P312 - P361 + P364