Pesticide reference standard, Ipronidazole-d3, VETRANAL(TM), analytical standard, Supelco®
Catalog # 34216-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:1015855-83-0
- Description:Ipronidazole-d3, VETRANAL(TM)
- M-Clarity quality level:MQ100
- Formula:C₇D₃H₈N₃O₂
- Description:Ipronidazole-d3, VETRANAL(TM)