Supelco® PESTANAL® Organic Reference Standard, Indaziflam
Catalog # 32096-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:950782-86-2
- Description:Indaziflam
- Storage conditions:2...30 °C
- Hazard class description:H410
- Shipping conditions:STD
- MDL No.:MFCD17167079
- M-Clarity quality level:MQ100
- Synonym(s):2-Amino-4-[(1R,2S)-2,6-dimethyl-indan-1-ylamino]-6-(1-fluoroethyl)-1,3,5-triazine; N-[(1R,2S)-2,3-Dihydro-2,6-dimethyl-1H-inden-1-yl]-6-[(1RS)-1-fluoroethyl]-1,3,5-triazine-2,4-diamine
- Formula:C₁₆H₂₀FN₅
- Molecular weight:301,36
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS09
Caution: P273 - P391 - P501