Supelco® PESTANAL® Organic Reference Standard, Imazaquin
37878-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:81335-37-7
- Description:Imazaquin
- Storage conditions:2...30 °C
- Hazard class description:H312
- Shipping conditions:STD
- MDL No.:MFCD00778521
- M-Clarity quality level:MQ100
- Synonym(s):2-(4-Isopropyl-4-methyl-5-oxo-4,5-dihydro-1H-imidazol-2-yl)-quinoline-3-carboxylic acid
- Formula:C₁₇H₁₇N₃O₃
- Molecular weight:311,34
- Flash point:Not applicable
- GHS signal: GHS07
: P280