Supelco® PESTANAL® Organic Reference Standard, Heptachlor Endo-Epoxide
Catalog # 35492-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:PESTANAL®, Analytical standard, isomer A
- CAS No.:28044-83-9
- Description:Heptachlor endo-epoxide
- Storage conditions:2...30 °C
- Hazard class description:H300 - H351 - H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD00872082
- M-Clarity quality level:MQ100
- Synonym(s):endo-1,4,5,6,7,8,8-Heptachloro-2,3-epoxy-4,7-methano-3a,4,7,7a-tetrahydroindane
- Formula:C₁₀H₅Cl₇O
- Molecular weight:389,32
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS08, GHS06, GHS09
Caution: P202 - P260 - P264 - P273 - P280 - P301 + P310