Supelco® PESTANAL® Organic Reference Standard, HCH
Catalog # 36756-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard, mixture of isomers (alpha:beta:gamma:delta=1:1:1:1)
- CAS No.:608-73-1
- Description:HCH
- Storage conditions:2...30 °C
- Hazard class description:H301 - H312 - H351 - H362 - H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD00003825
- M-Clarity quality level:MQ100
- Formula:C₆H₆Cl₆
- Molecular weight:290,83
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS08, GHS09
Caution: P260 - P263 - P273 - P280 - P301 + P310 - P302 + P352 + P312