Supelco® Organic Reference Standard, Glyceryl trinonadecanoate
52909-25MG
: Merck
:
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:26536-13-0
- Description:Glyceryl trinonadecanoate
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00056228
- M-Clarity quality level:MQ100
- Synonym(s):Trinonadecanoin
- Molecular weight:933,56