Order Entry
Slovakia
ContactUsLinkComponent
Supelco® Organic Reference Standard, Glyceryl trilinoleate
Supelco® Organic Reference Standard, Glyceryl trilinoleate
Catalog # 96373-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Glyceryl trilinoleate
Catalog # 96373-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    537-40-6
  • Description:
    Glyceryl trilinoleate
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H413
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00042909
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    1,2,3-Tri-(cis,cis-9,12-octadecadienoyl)glycerol; 1,2,3-Trilinoleoylglycerol; Glycerol trilinoleate; Trilinolein
  • Molecular weight:
    879,38
  • Density:
    0,925 g/ml at 20 °C (lit.)

Specifications

About this item

Caution: P273 - P501