Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Fluorodifen
Supelco® PESTANAL® Organic Reference Standard, Fluorodifen
Catalog # 45506-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Fluorodifen
Catalog # 45506-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    15457-05-3
  • Description:
    Fluorodifen
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H319 - H332 - H400
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055532
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₃H₇F₃N₂O₅
  • Molecular weight:
    328,20
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P273 - P304 + P340 + P312 - P305 + P351 + P338