Supelco® PESTANAL® Organic Reference Standard, Flumioxazin
32525-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:103361-09-7
- Description:Flumioxazin
- Storage conditions:2...30 °C
- Hazard class description:H361d - H410
- Shipping conditions:STD
- MDL No.:MFCD01754256
- M-Clarity quality level:MQ100
- Synonym(s):2-[7-Fluoro-3,4-dihydro-3-oxo-4-(2-propynyl)-2H-1,4-benzoxazin-6-yl]-4,5,6,7-tetrahydro-1H-isoindole-1,3(2H)-dione; Flumioxazine
- Formula:C₁₉H₁₅FN₂O₄
- Molecular weight:354,33
- Flash point:Not applicable
- GHS signal: GHS08, GHS09
: P202 - P273 - P280 - P308 + P313 - P391 - P405