Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Fipronil-(pyrazole-¹³C₃, cyano-¹³C)
Supelco® PESTANAL® Organic Reference Standard, Fipronil-(pyrazole-¹³C₃, cyano-¹³C)
Catalog # 79157-5MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Fipronil-(pyrazole-¹³C₃, cyano-¹³C)
Catalog # 79157-5MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Standards:
    PESTANAL®, analytical standard
  • Description:
    Fipronil-(pyrazole-¹³C₃, cyano-¹³C)
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H301 + H311 + H331 - H372 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD30738206
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    5-Amino-3-cyano-¹³C-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]- 4-[(trifluoromethyl)sulfinyl]pyrazole-¹³C₃; Fipronil-¹³C₄
  • Molecular weight:
    441,12

Specifications

About this item

  • GHS signal: GHS06, GHS08, GHS09
Caution: P273 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311 - P314