Supelco® PESTANAL® Organic Reference Standard, Fenoxaprop-P-Ethyl
36851-250MG
: Merck
:
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:71283-80-2
- Description:Fenoxaprop-P-ethyl
- Storage conditions:2...8 °C
- Hazard class description:H317 - H373 - H410
- Shipping conditions:STD
- MDL No.:MFCD01310637
- M-Clarity quality level:MQ100
- Synonym(s):Ethyl (R)-2-{4-[(6-chlorobenzoxazol-2-yl)oxy]phenoxy}propionate
- Formula:C₁₈H₁₆ClNO₅
- Molecular weight:361,78
- Storage temperature:2...8 °C
- Flash point:Not applicable
- GHS signal: GHS09
: P260 - P272 - P273 - P280 - P302 + P352 - P314