Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Famoxadone Solution
  33495-2ML-R
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Famoxadone Solution
  33495-2ML-R
 :  Merck
 :  

 

  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • CAS No.:
    131807-57-3
  • Description:
    Famoxadone solution
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD03427409
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    3-Anilino-5-methyl-5-(4-phenoxyphenyl)oxazolidine-2,4-dione
  • Formula:
    C₂₂H₁₈N₂O₄
  • Molecular weight:
    374,39
  • Storage temperature:
    2...8 °C
  • Flash point:
    2 °C

 

 

  • GHS signal: GHS02, GHS07
 : P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338