Supelco® PESTANAL® Organic Reference Standard, Etobenzanid
Catalog # 77065-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:79540-50-4
- Description:Etobenzanid
- Storage conditions:2...30 °C
- Hazard class description:H411
- Shipping conditions:STD
- MDL No.:MFCD01631156
- M-Clarity quality level:MQ100
- Synonym(s):2′,3′-Dichloro-4-ethoxymethoxybenzanilide; N-(2,3-Dichlorophenyl)-4-(ethoxymethoxy)benzamide
- Molecular weight:340,20
Specifications
About this item
- GHS signal: GHS09
Caution: P273