Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Ethiprole
Supelco® PESTANAL® Organic Reference Standard, Ethiprole
Catalog # 33976-100MG-R
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Ethiprole
Catalog # 33976-100MG-R
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    181587-01-9
  • Description:
    Ethiprole
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H332 - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD07366662
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    5-Amino-1-(2,6-dichloro-α,α,α-trifluoro-p-tolyl)-4-(ethylsulfinyl)pyrazole-3-carbonitrile
  • Formula:
    C₁₃H₉Cl₂F₃N₄OS
  • Molecular weight:
    397,20
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS08, GHS09
Caution: P260 - P271 - P273 - P304 + P340 + P312 - P314 - P391