Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Esfenvalerate
Supelco® PESTANAL® Organic Reference Standard, Esfenvalerate
Catalog # 46277-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Esfenvalerate
Catalog # 46277-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, analytical standard
  • CAS No.:
    66230-04-4
  • Description:
    Esfenvalerate
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H301 + H331 - H317 - H370 - H373 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00203302
  • M-Clarity quality level:
    MQ100
  • Molecular weight:
    419,90

Specifications

About this item

  • GHS signal: GHS06, GHS09
Caution: P260 - P273 - P280 - P301 + P310 - P302 + P352 - P304 + P340 + P311