Supelco® PESTANAL® Organic Reference Standard, EPTC
Catalog # 45469-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:759-94-4
- Description:EPTC
- Storage conditions:2...30 °C
- Hazard class description:H302 + H312 - H315 - H317 - H319 - H331 - H411
- Shipping conditions:STD
- MDL No.:MFCD00053780
- M-Clarity quality level:MQ100
- Synonym(s):S-Ethyl-N,N-dipropylthiocarbamate
- Formula:C₉H₁₉NOS
- Molecular weight:189,32
- Flash point:>100 °C
Specifications
About this item
- GHS signal: GHS07
Caution: P273 - P280 - P301 + P312 - P302 + P352 + P312 - P304 + P340 + P311 - P305 + P351 + P338