Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Endrin
Supelco® PESTANAL® Organic Reference Standard, Endrin
Catalog # 32014-100MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Endrin
Catalog # 32014-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    72-20-8
  • Description:
    Endrin
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H300 + H310 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00151158
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₂H₈Cl₆O
  • Molecular weight:
    380,91
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS06, GHS09
Caution: P262 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P310