Supelco® PESTANAL® Organic Reference Standard, Endosulfan Lactone
Catalog # 36675-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:3868-61-9
- Description:Endosulfan lactone
- Storage conditions:2...30 °C
- Hazard class description:H301
- Shipping conditions:STD
- MDL No.:MFCD00168050
- M-Clarity quality level:MQ100
- Formula:C₉H₄Cl₆O₂
- Molecular weight:356,84
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06
Caution: P301 + P310