Supelco® PESTANAL® Organic Reference Standard, Dimethylpropiothetin hydrochloride
80828-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:4337-33-1
- Description:Dimethylpropiothetin hydrochloride
- Storage conditions:2…8 °C
- Shipping conditions:STD
- MDL No.:MFCD00142888
- M-Clarity quality level:MQ100
- Synonym(s):DMSP; Dimethyl-3-propiothetin chloride; Dimethylsulfoniopropionate hydrochloride
- Molecular weight:170,66