Supelco® PESTANAL® Organic Reference Standard, Dimethametryn
Catalog # 45448-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:22936-75-0
- Description:Dimethametryn
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD00078649
- M-Clarity quality level:MQ100
- Formula:C₁₁H₂₁N₅S
- Molecular weight:255,38
- Flash point:Not applicable