Supelco® PESTANAL® Organic Reference Standard, Diflufenzopyr Sodium Salt
37916-100MG-R
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:109293-98-3
- Description:Diflufenzopyr sodium salt
- Storage conditions:2...30 °C
- Hazard class description:H332 - H412
- Shipping conditions:STD
- MDL No.:MFCD04974587
- M-Clarity quality level:MQ100
- Synonym(s):2-{1-(3,5-Difluorophenylaminocarbonylhydrazono)ethyl}-3-pyridinecarboxylic acid monosodium salt
- Formula:C₁₅H₁₁F₂N₄NaO₃
- Molecular weight:356,26
- Flash point:Not applicable
- GHS signal: GHS07
: P261 - P271 - P273 - P304 + P340 + P312 - P501