Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Difenoconazol
Supelco® PESTANAL® Organic Reference Standard, Difenoconazol
Catalog # 36531-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Difenoconazol
Catalog # 36531-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    119446-68-3
  • Description:
    Difenoconazol
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H302 - H319 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00144119
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₁₉H₁₇Cl₂N₃O₃
  • Molecular weight:
    406,26
  • Flash point:
    Not applicable

Specifications

About this item

  • GHS signal: GHS07, GHS09
Caution: P264 - P270 - P273 - P280 - P301 + P312 - P305 + P351 + P338