Supelco® PESTANAL® Organic Reference Standard, Dicrotophos
Catalog # 45305-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:141-66-2
- Description:Dicrotophos
- Storage conditions:2...8 °C
- Hazard class description:H300 + H310 + H330 - H410
- Shipping conditions:STD
- MDL No.:MFCD00072499
- M-Clarity quality level:MQ100
- Formula:C₈H₁₆NO₅P
- Molecular weight:237,19
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P260 - P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310