Supelco® PESTANAL® Organic Reference Standard, Dichlorvos-(dimethyl-d₆)
Catalog # 74867-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:203645-53-8
- Description:Dichlorvos-(dimethyl-d₆)
- Storage conditions:–20 °C
- Hazard class description:H300 + H310 + H330 - H317 - H400
- Shipping conditions:STD
- MDL No.:MFCD00209708
- M-Clarity quality level:MQ100
- Synonym(s):2,2-Dichlorovinyl di(methyl-d₃) phosphate; Dichlorvos-d₆
- Molecular weight:227,01
- Flash point:100 °C - Closed cup
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P260 - P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310