Supelco® PESTANAL® Organic Reference Standard, Desmethyl-Pirimicarb
Catalog # 33886-10MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:30614-22-3
- Description:Desmethyl-pirimicarb
- Storage conditions:2...8 °C
- Hazard class description:H300 - H315 - H319 - H335 - H400
- Shipping conditions:STD
- MDL No.:MFCD01940516
- M-Clarity quality level:MQ100
- Synonym(s):5,6-Dimethyl-2-methylamino-4-pyrimidinyl dimethylcarbamate
- Formula:C₁₀H₁₆N₄O₂
- Molecular weight:224,26
- Storage temperature:2...8 °C
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06, GHS09
Caution: P273 - P301 + P310 + P330 - P302 + P352 - P305 + P351 + P338