Supelco® PESTANAL® Organic Reference Standard, Desmethyl-Formamido-Pirimicarb
33887-10MG-R
: Merck
:
- Pk:10 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:27218-04-8
- Description:Desmethyl-formamido-pirimicarb
- Storage conditions:2...8 °C
- Hazard class description:H301 - H410
- Shipping conditions:STD
- MDL No.:MFCD08460514
- M-Clarity quality level:MQ100
- Synonym(s):5,6-Dimethyl-2-(N-formylmethylamino)-4-pyrimidinyl dimethylcarbamate
- Formula:C₁₁H₁₆N₄O₃
- Molecular weight:252,27
- Storage temperature:2...8 °C
- Flash point:Not applicable
- GHS signal: GHS06, GHS09
: P273 - P301 + P310 + P330