Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, Demeton-S-Methyl Solution
Catalog # 34234-2ML
Supplier:  Merck
CAS Number:  
undefined
Supelco® Organic Reference Standard, Demeton-S-Methyl Solution
Catalog # 34234-2ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    2 mL
  • Standards:
    Analytical standard
  • CAS No.:
    919-86-8
  • Description:
    Demeton-S-methyl solution
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H225 - H302 + H312 + H332 - H319
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055464
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    S-[2-(Ethylthio)ethyl] O,O-dimethyl phosphorothioate; Methyldemetone
  • Formula:
    C₆H₁₅O₃PS₂
  • Molecular weight:
    230,29
  • Storage temperature:
    2...8 °C
  • Flash point:
    2 °C

Specifications

About this item

  • GHS signal: GHS02, GHS07
Caution: P210 - P280 - P301 + P312 - P303 + P361 + P353 - P304 + P340 + P312 - P305 + P351 + P338