Order Entry
Slovakia
ContactUsLinkComponent
Supelco® PESTANAL® Organic Reference Standard, Deltamethrin
Supelco® PESTANAL® Organic Reference Standard, Deltamethrin
Catalog # 45423-250MG
Supplier:  Merck
CAS Number:  
undefined
Supelco® PESTANAL® Organic Reference Standard, Deltamethrin
Catalog # 45423-250MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    250 mg
  • Standards:
    PESTANAL®, Analytical standard
  • CAS No.:
    52918-63-5
  • Description:
    Deltamethrin
  • Storage conditions:
    2...30 °C
  • Hazard class description:
    H301 + H331 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00870122
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₂₂H₁₉Br₂NO₃
  • Molecular weight:
    505,20
  • Flash point:
    100 °C - closed cup

Specifications

About this item

  • GHS signal: GHS06, GHS09
Caution: P261 - P264 - P270 - P273 - P301 + P310 - P304 + P340 + P311