Supelco® PESTANAL® Organic Reference Standard, Cyprosulfamide
32522-100MG
: Merck
:
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:221667-31-8
- Description:Cyprosulfamide
- Storage conditions:2...30 °C
- Shipping conditions:STD
- MDL No.:MFCD15146679
- M-Clarity quality level:MQ100
- Synonym(s):N-[4-(Cyclopropylaminocarbonyl)phenylsulfonyl]-2-methoxybenzamide
- Formula:C₁₈H₁₈N₂O₅S
- Molecular weight:374,41