Supelco® PESTANAL® Organic Reference Standard, Cyprodinil
Catalog # 34389-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:121552-61-2
- Description:Cyprodinil
- Storage conditions:RT
- Hazard class description:H317 - H410
- Shipping conditions:STD
- MDL No.:MFCD01632330
- Synonym(s):4-Cyclopropyl-6-methyl-N-phenylpyrimidin-2-amine
- Formula:C₁₄H₁₅N₃
- Molecular weight:225,29
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07, GHS09
Caution: P261 - P272 - P273 - P280 - P302 + P352 - P333 + P313