Supelco® PESTANAL® Organic Reference Standard, Cycloate
Catalog # 45408-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:1134-23-2
- Description:Cycloate
- Storage conditions:2...30 °C
- Hazard class description:H331 - H411
- Shipping conditions:STD
- MDL No.:MFCD00055341
- M-Clarity quality level:MQ100
- Synonym(s):S-Ethyl N-cyclohexyl-N-ethylthiocarbamate
- Formula:C₁₁H₂₁NOS
- Molecular weight:215,36
- Flash point:>100 °C
Specifications
About this item
- GHS signal: GHS07
Caution: P261 - P271 - P273 - P304 + P340 + P311 - P391 - P403 + P233