Pesticide reference standard, Cyanuric acid-13C3, PESTANAL®, analytical standard, Supelco®
Catalog # 32679-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:201996-37-4
- Description:Cyanuric acid-13C3
- Melting point:>360 °C
- Formula:₁₃C₃H₃N₃O₃₃
- Description:Cyanuric acid-13C3