Order Entry
Slovakia
ContactUsLinkComponent
 
Supelco® PESTANAL® Organic Reference Standard, Cyanophos
  46279-25MG
 :  Merck
 :  
undefined
Supelco® PESTANAL® Organic Reference Standard, Cyanophos
  46279-25MG
 :  Merck
 :  

 

  • Pk:
    25 mg
  • Standards:
    PESTANAL®, analytical standard
  • CAS No.:
    2636-26-2
  • Description:
    Cyanophos
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H301 + H311 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055485
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    O-(4-Cyanophenyl) O,O-dimethyl phosphorothioate
  • Molecular weight:
    243,22
  • Flash point:
    >100 °C

 

 

  • GHS signal: GHS07, GHS09
 : P264 - P270 - P273 - P280 - P301 + P310 - P302 + P352 + P312