Supelco® PESTANAL® Organic Reference Standard, Clomazone
Catalog # 46120-100MG-R
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, analytical standard
- CAS No.:81777-89-1
- Description:Clomazone
- Storage conditions:2…8 °C
- Hazard class description:H302 + H332 - H410
- Shipping conditions:STD
- MDL No.:MFCD00072457
- M-Clarity quality level:MQ100
- Molecular weight:239,70
- Flash point:157 °C
Specifications
About this item
- GHS signal: GHS07
Caution: P261 - P264 - P271 - P273 - P301 + P312 - P304 + P340 + P312